3-Methyl-1,1-diphenylurea
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-3,3-diphenylurea
OR45151-25G
OR45151-5G
APOSOR45151-25GEA
132
EUR
APOSOR45151-25G
APOSOR45151-5G
3-Methyl-1,1-diphenylurea
3-Methyl-1,1-diphenylurea
Formel:
C₁₄H₁₄N₂O Molecular Weight: 226,274 g/mol Schmelzpunkt: 172…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043722 CAS-Nummer: 13114-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...