Ethyl 6-chlorobenzofuran-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR45160-1G
OR45160-250MG
APOSOR45160-1GEA
998
EUR
APOSOR45160-1G
APOSOR45160-250MG
Ethyl 6-chlorobenzofuran-2-carboxylate
Ethyl 6-chlorobenzofuran-2-carboxylate
Formel:
C₁₁H₉ClO₃ Molecular Weight: 224,64 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28987315 CAS-Nummer: 74222-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...