6-Chlorobenzofuran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR45162-1G OR45162-250MG
APOSOR45162-1GEA 102 EUR
APOSOR45162-1G APOSOR45162-250MG
6-Chlorobenzofuran-2-carboxylic acid
6-Chlorobenzofuran-2-carboxylic acid
Formel: C₉H₅ClO₃
Molecular Weight: 196,587 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20528041
CAS-Nummer: 442125-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR