7-Bromo-5-methoxybenzofuran-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR45167-1G
OR45167-250MG
APOSOR45167-1GEA
145
EUR
APOSOR45167-1G
APOSOR45167-250MG
7-Bromo-5-methoxybenzofuran-2-carboxylic acid
7-Bromo-5-methoxybenzofuran-2-carboxylic acid
Formel:
C₁₀H₇BrO₄ Molecular Weight: 271,064 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22574014 CAS-Nummer: 875830-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...