5-Bromo-4-(cyclopropyl)thiophene-2-carboxaldehyde

Lieferant: Apollo Scientific

OR45200-1G OR45200-250MG
APOSOR45200-1GEA 696 EUR
APOSOR45200-1G APOSOR45200-250MG
5-Bromo-4-(cyclopropyl)thiophene-2-carboxaldehyde
5-Bromo-4-(cyclopropyl)thiophene-2-carboxaldehyde
Formel: C₈H₇BrOS
Molecular Weight: 231,11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32219463
CAS-Nummer: 2301169-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR