Boc-5-Bromo-L-tryptophan 97%
Lieferant: Apollo Scientific
OR452006-1G
OR452006-250MG
OR452006-500MG
APOSOR452006-1GEA
654
EUR
APOSOR452006-1G
APOSOR452006-250MG
APOSOR452006-500MG
Boc-5-Bromo-L-tryptophan 97%
Boc-5-Bromo-L-tryptophan
Formel:
C₁₆H₁₉BrN₂O₄ Molecular Weight: 383,237 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09264326 CAS-Nummer: 75816-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...