4-(Cyclopropyl)-2-(hydroxymethyl)thiophene
Lieferant: Apollo Scientific
OR45201-1G
OR45201-250MG
APOSOR45201-1GEA
696
EUR
APOSOR45201-1G
APOSOR45201-250MG
4-(Cyclopropyl)-2-(hydroxymethyl)thiophene
4-(Cyclopropyl)-2-(hydroxymethyl)thiophene
Formel:
C₈H₁₀OS Molecular Weight: 154,229 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD32219464 CAS-Nummer: 2416262-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...