Boc-(S)-3-Cyclobutylalanine 97%

Lieferant: Apollo Scientific

OR452011-1G OR452011-250MG OR452011-500MG
APOSOR452011-1GEA 522 EUR
APOSOR452011-1G APOSOR452011-250MG APOSOR452011-500MG
Boc-(S)-3-Cyclobutylalanine 97%
Boc-(S)-3-Cyclobutylalanine
Formel: C₁₂H₂₁NO₄
Molecular Weight: 243,299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840392
CAS-Nummer: 478183-60-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR