2,3-Dichloro-DL-phenylalanine 97%
Lieferant: Apollo Scientific
OR452014-1G
OR452014-500MG
OR452014-5G
APOSOR452014-1GEA
197
EUR
APOSOR452014-1G
APOSOR452014-500MG
APOSOR452014-5G
2,3-Dichloro-DL-phenylalanine 97%
2,3-Dichloro-DL-phenylalanine
Formel:
C₉H₉Cl₂NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318243 CAS-Nummer: 110300-04-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...