3,4-Dimethyl-L-phenylalanine 97%
Lieferant: Apollo Scientific
OR452017-1G
OR452017-500MG
APOSOR452017-1GEA
522
EUR
APOSOR452017-1G
APOSOR452017-500MG
3,4-Dimethyl-L-phenylalanine 97%
3,4-Dimethyl-L-phenylalanine
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,242 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09264360 CAS-Nummer: 142995-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...