5-Methyl-L-tryptophan 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-2-Amino-3-(5-methyl-1H-indol-3-yl)propanoic acid, H-Trp(5-Me)-OH
OR452022-1G
OR452022-250MG
APOSOR452022-1GEA
607
EUR
APOSOR452022-1G
APOSOR452022-250MG
5-Methyl-L-tryptophan 97%
5-Methyl-L-tryptophan
Formel:
C₁₂H₁₄N₂O₂ Molecular Weight: 218,252 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01677297 CAS-Nummer: 154-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...