4-Bromo-D-tryptophan 97%

Lieferant: Apollo Scientific

OR452028-1G OR452028-250MG OR452028-500MG
APOSOR452028-1GEA 980 EUR
APOSOR452028-1G APOSOR452028-250MG APOSOR452028-500MG
4-Bromo-D-tryptophan 97%
4-Bromo-D-tryptophan
Formel: C₁₁H₁₁BrN₂O₂
Molecular Weight: 283,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10565750
CAS-Nummer: 219943-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR