5-Bromo-4-(cyclopropyl)thiophene-2-carbonitrile

Lieferant: Apollo Scientific

OR45203-1G OR45203-250MG
APOSOR45203-1GEA 696 EUR
APOSOR45203-1G APOSOR45203-250MG
5-Bromo-4-(cyclopropyl)thiophene-2-carbonitrile
5-Bromo-4-(cyclopropyl)thiophene-2-carbonitrile
Formel: C₈H₆BrNS
Molecular Weight: 228,109 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32219465
CAS-Nummer: 2301170-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR