4-Chloro-L-tryptophan 97%

Lieferant: Apollo Scientific

Synonyme: H-Trp(4-Cl)-OH

OR452030-1G OR452030-250MG
APOSOR452030-1GEA 340 EUR
APOSOR452030-1G APOSOR452030-250MG
4-Chloro-L-tryptophan 97%
4-Chloro-L-tryptophan
Formel: C₁₁H₁₁ClN₂O₂
Molecular Weight: 238,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797630
CAS-Nummer: 52448-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR