4-Chloro-D-tryptophan 97%
Lieferant: Apollo Scientific
OR452031-1G
OR452031-250MG
OR452031-500MG
APOSOR452031-1GEA
980
EUR
APOSOR452031-1G
APOSOR452031-250MG
APOSOR452031-500MG
4-Chloro-D-tryptophan 97%
4-Chloro-D-tryptophan
Formel:
C₁₁H₁₁ClN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10565751 CAS-Nummer: 27542-41-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...