(R)-2-Amino-3-biphenyl-3-yl-propionic acid 97%

Lieferant: Apollo Scientific

OR452034-1G OR452034-250MG OR452034-500MG
APOSOR452034-1GEA 1050 EUR
APOSOR452034-1G APOSOR452034-250MG APOSOR452034-500MG
(R)-2-Amino-3-biphenyl-3-yl-propionic acid 97%
(R)-2-Amino-3-biphenyl-3-yl-propionic acid
Formel: C₁₅H₁₅NO₂
Molecular Weight: 241,285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07372094
CAS-Nummer: 164172-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR