Boc-(S)-2-Amino-3-biphenyl-3-yl-propionic acid 97%
Lieferant: Apollo Scientific
OR452035-1G
OR452035-250MG
OR452035-500MG
APOSOR452035-1GEA
1050
EUR
APOSOR452035-1G
APOSOR452035-250MG
APOSOR452035-500MG
Boc-(S)-2-Amino-3-biphenyl-3-yl-propionic acid 97%
Boc-(S)-2-Amino-3-biphenyl-3-yl-propionic acid
Formel:
C₂₀H₂₃NO₄ Molecular Weight: 341,401 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07372097 CAS-Nummer: 608528-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...