N-Fmoc-3-hydroxy-D-phenylalanine 97%
Lieferant: Apollo Scientific
OR452044-1G
OR452044-250MG
OR452044-500MG
APOSOR452044-1GEA
654
EUR
APOSOR452044-1G
APOSOR452044-250MG
APOSOR452044-500MG
N-Fmoc-3-hydroxy-D-phenylalanine 97%
N-Fmoc-3-hydroxy-D-phenylalanine
Formel:
C₂₄H₂₁NO₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371960 CAS-Nummer: 1217724-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...