2,3-Dichloro-D-phenylalanine 97%

Lieferant: Apollo Scientific

OR452047-1G OR452047-500MG
APOSOR452047-1GEA 393 EUR
APOSOR452047-1G APOSOR452047-500MG
2,3-Dichloro-D-phenylalanine 97%
2,3-Dichloro-D-phenylalanine
Formel: C₉H₉Cl₂NO₂
Molecular Weight: 234,079 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07372010
CAS-Nummer: 1241677-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR