(S)-4-Nitro-homophenylalanine 97%

Lieferant: Apollo Scientific

OR452051-1G OR452051-250MG OR452051-500MG
APOSOR452051-1GEA 980 EUR
APOSOR452051-1G APOSOR452051-250MG APOSOR452051-500MG
(S)-4-Nitro-homophenylalanine 97%
(S)-4-Nitro-homophenylalanine
Formel: C₁₀H₁₂N₂O₄
Molecular Weight: 224.213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12546235
CAS-Nummer: 705919-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR