6-Methyl-L-tryptophan 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-2-Amino-3-(6-methyl-1H-indol-3-yl)propanoic acid, H-Trp(6-Me)-OH
OR452052-250MG
APOSOR452052-250MGEA
372
EUR
APOSOR452052-250MG
6-Methyl-L-tryptophan 97%
6-Methyl-L-tryptophan
Formel:
C₁₂H₁₄N₂O₂ Molecular Weight: 218,252 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797708 CAS-Nummer: 33468-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...