1,1-Bis(hydroxymethyl)cyclopentane 97%

Lieferant: Apollo Scientific

Synonyme: Cyclopentane-1,1-diyldimethanol

OR452056-1G OR452056-5G
APOSOR452056-1GEA 162 EUR
APOSOR452056-1G APOSOR452056-5G
1,1-Bis(hydroxymethyl)cyclopentane 97%
1,1-Bis(hydroxymethyl)cyclopentane
Formel: C₇H₁₄O₂
Molecular Weight: 130,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463707
CAS-Nummer: 5763-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR