2-(3,5-Dichlorophenoxy)acetic acid 97%
Lieferant: Apollo Scientific
OR452058-1G
OR452058-5G
APOSOR452058-1GEA
340
EUR
APOSOR452058-1G
APOSOR452058-5G
2-(3,5-Dichlorophenoxy)acetic acid 97%
2-(3,5-Dichlorophenoxy)acetic acid
Formel:
C₈H₆Cl₂O₃ Molecular Weight: 221.037 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037337 CAS-Nummer: 587-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...