5-(Cyclopropyl)thiophene-3-carboxylic acid
Lieferant: Apollo Scientific
OR45206-1G
OR45206-250MG
OR45206-5G
APOSOR45206-1GEA
0
EUR
APOSOR45206-1G
APOSOR45206-250MG
APOSOR45206-5G
5-(Cyclopropyl)thiophene-3-carboxylic acid
5-(Cyclopropyl)thiophene-3-carboxylic acid
Formel:
C₈H₈O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD23750170 CAS-Nummer: 1518721-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...