2-Morpholineacetic acid hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme: Morpholin-2-ylacetic acid hydrochloride
OR452078-1G OR452078-250MG OR452078-500MG
APOSOR452078-1GEA 654 EUR
APOSOR452078-1G APOSOR452078-250MG APOSOR452078-500MG
2-Morpholineacetic acid hydrochloride 97%
2-Morpholineacetic acid hydrochloride
Formel: C₆H₁₁NO₃·ClH
Molecular Weight: 181.617 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779523
CAS-Nummer: 1187929-25-4