Boc-N-methyl-D-norleucine 97%

Lieferant: Apollo Scientific

OR452086-1G OR452086-500MG OR452086-5G
APOSOR452086-1GEA 281 EUR
APOSOR452086-1G APOSOR452086-500MG APOSOR452086-5G
Boc-N-methyl-D-norleucine 97%
Boc-N-methyl-D-norleucine
Formel: C₁₂H₂₃NO₄
Molecular Weight: 245,315 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366766
CAS-Nummer: 161450-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR