Boc-D-methionine sulphone 97%

Lieferant: Apollo Scientific

OR452087-1G OR452087-500MG
APOSOR452087-1GEA 393 EUR
APOSOR452087-1G APOSOR452087-500MG
Boc-D-methionine sulphone 97%
Boc-D-methionine sulphone
Formel: C₁₀H₁₉NO₆S
Molecular Weight: 281,326 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21363159
CAS-Nummer: 74086-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR