5-(Cyclopropyl)thiophene-2-carbonitrile

Lieferant: Apollo Scientific

OR45209-1G OR45209-250MG
APOSOR45209-1GEA 696 EUR
APOSOR45209-1G APOSOR45209-250MG
5-(Cyclopropyl)thiophene-2-carbonitrile
5-(Cyclopropyl)thiophene-2-carbonitrile
Formel: C₈H₇NS
Molecular Weight: 149,213 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28059614
CAS-Nummer: 1823928-25-1
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR