N-Fmoc-(R)-2-(methylamino)butyric acid 97%

Lieferant: Apollo Scientific

OR452091-1G OR452091-500MG
APOSOR452091-1GEA 522 EUR
APOSOR452091-1G APOSOR452091-500MG
N-Fmoc-(R)-2-(methylamino)butyric acid 97%
N-Fmoc-(R)-2-(methylamino)butyric acid
Formel: C₂₀H₂₁NO₄
Molecular Weight: 339,385 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22380683
CAS-Nummer: 1210830-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR