Fmoc-2-aminobutyric acid 97%
Lieferant: Apollo Scientific
Synonyme:
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)butanoic acid, Fmoc-DL-Abu-OH
OR452095-25G
OR452095-5G
APOSOR452095-25GEA
172
EUR
APOSOR452095-25G
APOSOR452095-5G
Fmoc-2-aminobutyric acid 97%
Fmoc-2-aminobutyric acid
Formel:
C₁₉H₁₉NO₄ Molecular Weight: 325,358 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270362 CAS-Nummer: 174879-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...