N-Boc-(S)-2-(methylamino)butyric acid 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-2-((tert-Butoxycarbonyl)(methyl)amino)butanoic acid
OR452097-1G
OR452097-5G
APOSOR452097-1GEA
201
EUR
APOSOR452097-1G
APOSOR452097-5G
N-Boc-(S)-2-(methylamino)butyric acid 97%
N-Boc-(S)-2-(methylamino)butyric acid
Formel:
C₁₀H₁₉NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD17215563 CAS-Nummer: 101759-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...