N-Methyl-2-pyrrolidinecarboxamide hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-2-pyrrolidinecarboxamide HCl

OR452102-1G OR452102-500MG
APOSOR452102-1GEA 445 EUR
APOSOR452102-1G APOSOR452102-500MG
N-Methyl-2-pyrrolidinecarboxamide hydrochloride 97%
N-Methyl-2-pyrrolidinecarboxamide hydrochloride
Formel: C₆H₁₂N₂O·ClH
Molecular Weight: 164,633 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858066
CAS-Nummer: 859537-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR