3-Phenyl-3-pyrrolidinol hydrochloride 97%

Lieferant: Apollo Scientific

OR452104-1G OR452104-250MG OR452104-500MG
APOSOR452104-1GEA 783 EUR
APOSOR452104-1G APOSOR452104-250MG APOSOR452104-500MG
3-Phenyl-3-pyrrolidinol hydrochloride 97%
3-Phenyl-3-pyrrolidinol hydrochloride
Formel: C₁₀H₁₃NO·ClH
Molecular Weight: 199,677 g/mol
Schmelzpunkt: 145…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01687413
CAS-Nummer: 1488-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR