1-Fmoc-2-(hydroxymethyl)piperidine 97%

Lieferant: Apollo Scientific

OR452109-1G OR452109-500MG OR452109-5G
APOSOR452109-1GEA 261 EUR
APOSOR452109-1G APOSOR452109-500MG APOSOR452109-5G
1-Fmoc-2-(hydroxymethyl)piperidine 97%
1-Fmoc-2-(hydroxymethyl)piperidine
Formel: C₂₁H₂₃NO₃
Molecular Weight: 337.412 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04974380
CAS-Nummer: 170867-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR