1-Boc-4-cyano-4-benzyl-piperidine 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-benzyl-4-cyanopiperidine-1-carboxylate

OR452119-1G OR452119-250MG
APOSOR452119-1GEA 962 EUR
APOSOR452119-1G APOSOR452119-250MG
1-Boc-4-cyano-4-benzyl-piperidine 97%
1-Boc-4-cyano-4-benzyl-piperidine
Formel: C₁₈H₂₄N₂O₂
Molecular Weight: 300,395 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11849801
CAS-Nummer: 906329-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR