3-Phenyl-3-piperidinol hydrochloride 97%

Lieferant: Apollo Scientific

OR452120-1G OR452120-250MG OR452120-500MG
APOSOR452120-1GEA 654 EUR
APOSOR452120-1G APOSOR452120-250MG APOSOR452120-500MG
3-Phenyl-3-piperidinol hydrochloride 97%
3-Phenyl-3-piperidinol hydrochloride
Formel: C₁₁H₁₅NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD29050460
CAS-Nummer: 105558-52-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR