4-Methyl-1-benzyl-4-piperidinol hydrochloride 97%

Lieferant: Apollo Scientific

OR452126-1G OR452126-500MG OR452126-5G
APOSOR452126-1GEA 328 EUR
APOSOR452126-1G APOSOR452126-500MG APOSOR452126-5G
4-Methyl-1-benzyl-4-piperidinol hydrochloride 97%
4-Methyl-1-benzyl-4-piperidinol hydrochloride
Formel: C₁₃H₁₉NO·ClH
Molecular Weight: 241,757 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14582893
CAS-Nummer: 80676-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR