1-Fmoc-4-(hydroxymethyl)piperidine 97%

Lieferant: Apollo Scientific

OR452130-1G OR452130-500MG OR452130-5G
APOSOR452130-1GEA 219 EUR
APOSOR452130-1G APOSOR452130-500MG APOSOR452130-5G
1-Fmoc-4-(hydroxymethyl)piperidine 97%
1-Fmoc-4-(hydroxymethyl)piperidine
Formel: C₂₁H₂₃NO₃
Molecular Weight: 337,412 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02094489
CAS-Nummer: 1072502-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR