1-Boc-4-(4-methoxyphenyl)-4-carboxypiperidine 97%

Lieferant: Apollo Scientific

OR452132-1G OR452132-250MG
APOSOR452132-1GEA 661 EUR
APOSOR452132-1G APOSOR452132-250MG
1-Boc-4-(4-methoxyphenyl)-4-carboxypiperidine 97%
1-Boc-4-(4-methoxyphenyl)-4-carboxypiperidine
Formel: C₁₈H₂₅NO₅
Molecular Weight: 335,395 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11223621
CAS-Nummer: 1158750-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR