3-Methoxybenzenepropanol 97%

Lieferant: Apollo Scientific

Synonyme: 3-(3-Methoxyphenyl)propan-1-ol

OR452137-1G
APOSOR452137-1GEA 307 EUR
APOSOR452137-1G
3-Methoxybenzenepropanol 97%
3-Methoxybenzenepropanol
Formel: C₁₀H₁₄O₂
Molecular Weight: 166,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798002
CAS-Nummer: 7252-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR