1-(3-Ethynylphenyl)-ethanone 97%

Lieferant: Apollo Scientific

OR452138-1G OR452138-500MG
APOSOR452138-1GEA 480 EUR
APOSOR452138-1G APOSOR452138-500MG
1-(3-Ethynylphenyl)-ethanone 97%
1-(3-Ethynylphenyl)-ethanone
Formel: C₁₀H₈O
Molecular Weight: 144,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04974024
CAS-Nummer: 139697-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR