2-Amino-2-(2-methylphenyl)ethan-1-ol 97%

Lieferant: Apollo Scientific

OR452139-1G OR452139-250MG OR452139-500MG
APOSOR452139-1GEA 870 EUR
APOSOR452139-1G APOSOR452139-250MG APOSOR452139-500MG
2-Amino-2-(2-methylphenyl)ethan-1-ol 97%
2-Amino-2-(2-methylphenyl)ethan-1-ol
Formel: C₉H₁₃NO
Molecular Weight: 151.206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11213642
CAS-Nummer: 1094627-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR