Cbz-DL-Alaninol 97%
Lieferant: Apollo Scientific
OR452145-1G
OR452145-25G
OR452145-5G
APOSOR452145-1GEA
136
EUR
APOSOR452145-1G
APOSOR452145-25G
APOSOR452145-5G
Cbz-DL-Alaninol 97%
Cbz-DL-Alaninol
Formel:
C₁₁H₁₅NO₃ Molecular Weight: 209,242 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12180250 CAS-Nummer: 87905-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...