Cbz-(S)-2-phenylglycinol 97%
Lieferant: Apollo Scientific
OR452146-1G
OR452146-25G
OR452146-5G
APOSOR452146-1GEA
139
EUR
APOSOR452146-1G
APOSOR452146-25G
APOSOR452146-5G
Cbz-(S)-2-phenylglycinol 97%
Cbz-(S)-2-phenylglycinol
Formel:
C₁₆H₁₇NO₃ Molecular Weight: 271.311 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792493 CAS-Nummer: 130406-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...