Cbz-(R)-2-phenylglycinol 97%

Lieferant: Apollo Scientific

OR452147-1G OR452147-25G OR452147-5G
APOSOR452147-1GEA 139 EUR
APOSOR452147-1G APOSOR452147-25G APOSOR452147-5G
Cbz-(R)-2-phenylglycinol 97%
Cbz-(R)-2-phenylglycinol
Formel: C₁₆H₁₇NO₃
Molecular Weight: 271,311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00792494
CAS-Nummer: 120666-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR