Cbz-L-Valinol 97%
Lieferant: Apollo Scientific
Synonyme:
Cbz-Val-ol
OR452148-1G
OR452148-5G
APOSOR452148-1GEA
108
EUR
APOSOR452148-1G
APOSOR452148-5G
Cbz-L-Valinol 97%
Cbz-L-Valinol
Formel:
C₁₃H₁₉NO₃ Molecular Weight: 237,295 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672539 CAS-Nummer: 6216-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...