(S)-b-Amino-4-chloro-benzeneethanol 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-2-Amino-2-(4-chlorophenyl)ethanol
OR452154-1G
OR452154-250MG
APOSOR452154-1GEA
139
EUR
APOSOR452154-1G
APOSOR452154-250MG
(S)-b-Amino-4-chloro-benzeneethanol 97%
(S)-b-Amino-4-chloro-benzeneethanol
Formel:
C₈H₁₀ClNO Molecular Weight: 171,624 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09253716 CAS-Nummer: 191109-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...