N-Boc-N-methyl-2-bromobenzylamine 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2-bromobenzyl(methyl)carbamate

OR452165-1G OR452165-5G
APOSOR452165-1GEA 322 EUR
APOSOR452165-1G APOSOR452165-5G
N-Boc-N-methyl-2-bromobenzylamine 97%
N-Boc-N-methyl-2-bromobenzylamine
Formel: C₁₃H₁₈BrNO₂
Molecular Weight: 300,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09701231
CAS-Nummer: 954238-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR