N-Boc-N-methyl-3-bromobenzylamine 97%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 3-bromobenzyl(methyl)carbamate
OR452166-1G
OR452166-5G
APOSOR452166-1GEA
151
EUR
APOSOR452166-1G
APOSOR452166-5G
N-Boc-N-methyl-3-bromobenzylamine 97%
N-Boc-N-methyl-3-bromobenzylamine
Formel:
C₁₃H₁₈BrNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09701230 CAS-Nummer: 317358-61-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...