b-Amino-4-chlorobenzeneethanol 97%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-2-(4-chlorophenyl)ethanol

OR452167-1G OR452167-250MG
APOSOR452167-1GEA 149 EUR
APOSOR452167-1G APOSOR452167-250MG
b-Amino-4-chlorobenzeneethanol 97%
b-Amino-4-chlorobenzeneethanol
Formel: C₈H₁₀ClNO
Molecular Weight: 171.624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09759235
CAS-Nummer: 179811-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR